5-Methylchinoxalin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS7007G1 AS7007G5
ACTIAS7007G1EA 0 CHF
ACTIAS7007G1 ACTIAS7007G5
5-Methylchinoxalin
5-Methylquinoxaline
Formel: C₉H₈N₂
MW: 144,18 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00012335
CAS-Nummer: 13708-12-8
EINECS: 237-246-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR