3-Methyl-2-pyridon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methyl-2-pyridinone, 2-Hydroxy-3-methylpyridine, 3-Methylpyridin-2(1H)-one

AS32203G1 AS32203G10 AS32203G5
ACTIAS32203G1EA 0 CHF
ACTIAS32203G1 ACTIAS32203G10 ACTIAS32203G5
3-Methyl-2-pyridon
3-Methyl-2-pyridon
Formel: C₆H₇NO
MW: 109,13 g/mol
MDL Number: MFCD00039704
CAS-Nummer: 1003-56-1
EINECS: 213-710-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR