4-Hydroxyindol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
AS50142G100
AS50142G5
ACTIAS50142G100EA
0
CHF
ACTIAS50142G100
ACTIAS50142G5
4-Hydroxyindol
4-Hydroxyindol
Formel:
C₈H₇NO MW: 133,15 g/mol Siedepunkt: 100 °C (0,01 mmHg) Melting Pt: 102…104 °C |
MDL Number:
MFCD00005667 CAS-Nummer: 2380-94-1 EINECS: 219-177-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...