2-Amino-5-methyl-3-nitropyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Amino-5-nitro-3-picoline, 2-Amino-3-nitro-5-picoline, 5-Methyl-3-nitro-2-pyridylamine, 5-Methyl-3-nitropyridin-2-amine
AS34901G1
AS34901G10
AS34901G5
ACTIAS34901G1EA
0
CHF
ACTIAS34901G1
ACTIAS34901G10
ACTIAS34901G5
2-Amino-5-methyl-3-nitropyridin
2-Amino-5-methyl-3-nitropyridin
Formel:
C₆H₇N₃O₂ MW: 153,14 g/mol |
MDL Number:
MFCD00047442 CAS-Nummer: 7598-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...