2-Amino-3-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS20527G1 AS20527G25 AS20527G5
ACTIAS20527G1EA 0 CHF
ACTIAS20527G1 ACTIAS20527G25 ACTIAS20527G5
2-Amino-3-iodpyridin
2-Amino-3-iodopyridine
Formel: C₅H₅IN₂
MW: 220,01 g/mol
MDL Number: MFCD01651885
CAS-Nummer: 104830-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR