2-Fluor-5-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2-Fluoro-5-pyridinol, 6-Fluoropyridin-3-ol

AS30504G1 AS30504G5
ACTIAS30504G1EA 0 CHF
ACTIAS30504G1 ACTIAS30504G5
2-Fluor-5-hydroxypyridin
2-Fluor-5-hydroxypyridin
Formel: C₅H₄FNO
MW: 113,09 g/mol
MDL Number: MFCD01075040
CAS-Nummer: 55758-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR