3-Nitro-5-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS7089G1 AS7089G5
ACTIAS7089G1EA 0 CHF
ACTIAS7089G1 ACTIAS7089G5
3-Nitro-5-(trifluormethyl)anilin
3-Nitro-5-(trifluormethyl)anilin
Formel: C₇H₅F₃N₂O₂
MW: 206,12 g/mol
MDL Number: MFCD00061229
CAS-Nummer: 401-94-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR