2-Fluor-4-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS50173G1 AS50173G5
ACTIAS50173G1EA 18.74 CHF
ACTIAS50173G1 ACTIAS50173G5
2-Fluor-4-iodpyridin
2-Fluor-4-iodpyridin
Formel: C₅H₃FIN
MW: 222,99 g/mol
MDL Number: MFCD03092926
CAS-Nummer: 22282-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR