3-(Trifluormethoxy)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33301G1 AS33301G5
ACTIAS33301G1EA 0 CHF
ACTIAS33301G1 ACTIAS33301G5
3-(Trifluormethoxy)benzonitril
3-(Trifluormethoxy)benzonitril
Formel: C₈H₄F₃NO
MW: 187,12 g/mol
Siedepunkt: 236 °C (1013 hPa)
Melting Pt: 37 °C
MDL Number: MFCD00040959
CAS-Nummer: 52771-22-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR