5,6-Difluorindol

Lieferant: ACTIVATE SCIENTIFIC

AS31391G1 AS31391G5
ACTIAS31391G1EA 0 CHF
ACTIAS31391G1 ACTIAS31391G5
5,6-Difluorindol
5,6-Difluorindol
Formel: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD01075212
CAS-Nummer: 169674-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR