2,3,5-Trifluorpyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS33308G1
AS33308G5
ACTIAS33308G1EA
0
CHF
ACTIAS33308G1
ACTIAS33308G5
2,3,5-Trifluorpyridin
2,3,5-Trifluoropyridine
Formel:
C₅H₂F₃N MW: 133.07 g/mol Siedepunkt: 102 Dichte: 1.499 |
MDL Number:
MFCD03001162 CAS-Nummer: 76469-41-5 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...