(3-Nitrophenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS34246G1 AS34246G10 AS34246G5
ACTIAS34246G1EA 0 CHF
ACTIAS34246G1 ACTIAS34246G10 ACTIAS34246G5
(3-Nitrophenyl)essigsäure
(3-Nitrophenyl)essigsäure
Formel: (C₆H₄(NO₂))CH₂COOH
MW: 181,15 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00007278
CAS-Nummer: 1877-73-2
EINECS: 217-512-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR