5-Hydroxy-L-tryptophan
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
(S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, 5-Hydroxyl-L-tryptophan, (-)-5-Hydroxy-L-Tryptophane, L-5-Hydroxytryptophan, Oxitriptan, H-Trp(5-OH)-OH
AS33285G1
AS33285G5
ACTIAS33285G1EA
0
CHF
ACTIAS33285G1
ACTIAS33285G5
5-Hydroxy-L-tryptophan
5-Hydroxy-L-tryptophan
Formel:
C₁₁H₁₂N₂O₃ Melting Pt: 273 °C Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00064341 CAS-Nummer: 4350-09-8 EINECS: 224-411-1 UN: 2811 ADR: 6.1,III Merck Index: 12,04895 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...