5-Methoxyanthranilsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS25157G25 AS25157G5
ACTIAS25157G25EA 0 CHF
ACTIAS25157G25 ACTIAS25157G5
5-Methoxyanthranilsäure
5-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00016509
CAS-Nummer: 6705-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR