2-Naphthylessigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

AS21961G100 AS21961G25
ACTIAS21961G100EA 0 CHF
ACTIAS21961G100 ACTIAS21961G25
2-Naphthylessigsäure
2-Naphthylessigsäure
Formel: C₁₂H₁₀O₂
MW: 186.21 g/mol
Siedepunkt: 352
Melting Pt: 139…142 °C
Dichte: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00004126
CAS-Nummer: 581-96-4
EINECS: 209-475-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR