2-Chlor-4-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS50157G100
AS50157G5
ACTIAS50157G100EA
0
CHF
ACTIAS50157G100
ACTIAS50157G5
2-Chlor-4-iodpyridin
2-Chlor-4-iodpyridin
Formel:
C₅H₃ClIN MW: 239,44 g/mol |
MDL Number:
MFCD01861983 CAS-Nummer: 153034-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...