3-Chlor-2-iodtoluol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 1-Chloro-2-iodo-3-methylbenzene

AS33831G1 AS33831G10 AS33831G5
ACTIAS33831G1EA 0 CHF
ACTIAS33831G1 ACTIAS33831G10 ACTIAS33831G5
3-Chlor-2-iodtoluol
3-Chlor-2-iodtoluol
Formel: C₇H₆ClI
MW: 252,48 g/mol
MDL Number: MFCD00051756
CAS-Nummer: 5100-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR