3-Chlor-2-fluoranisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Chloro-2-fluoro-3-methoxybenzene

AS34649G1 AS34649G10 AS34649G5
ACTIAS34649G1EA 0 CHF
ACTIAS34649G1 ACTIAS34649G10 ACTIAS34649G5
3-Chlor-2-fluoranisol
3-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
MW: 160,58 g/mol
MDL Number: MFCD01631570
CAS-Nummer: 261762-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR