2,6-Dichlor-4-(trifluormethyl)anilin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS33529G10
AS33529G100
AS33529G5
ACTIAS33529G10EA
0
CHF
ACTIAS33529G10
ACTIAS33529G100
ACTIAS33529G5
2,6-Dichlor-4-(trifluormethyl)anilin
2,6-Dichlor-4-(trifluormethyl)anilin
Formel:
C₇H₄Cl₂F₃N Siedepunkt: 60…62 °C (1 mmHg) Melting Pt: 32…35 °C Dichte: 1,532 g/cm³ (20 °C) |
MDL Number:
MFCD00052918 CAS-Nummer: 24279-39-8 EINECS: 416-430-0 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...