3-Chlor-6-methylpyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS50532G1 AS50532G100 AS50532G5
ACTIAS50532G1EA 0 CHF
ACTIAS50532G1 ACTIAS50532G100 ACTIAS50532G5
3-Chlor-6-methylpyridazin
3-Chloro-6-methylpyridazine
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
MDL Number: MFCD00052905
CAS-Nummer: 1121-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR