3,4,5-Trichlorpyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS32181G1 AS32181G5
ACTIAS32181G1EA 0 CHF
ACTIAS32181G1 ACTIAS32181G5
3,4,5-Trichlorpyridazin
3,4,5-Trichloropyridazine
Formel: C₄HCl₃N₂
MDL Number: MFCD00006464
CAS-Nummer: 14161-11-6
EINECS: 238-006-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR