3-Cyanbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-Formylbenzonitrile

AS23457G100 AS23457G25
ACTIAS23457G100EA 0 CHF
ACTIAS23457G100 ACTIAS23457G25
3-Cyanbenzaldehyd
3-Cyanbenzaldehyd
Formel: C₈H₅NO
MW: 131,13 g/mol
Siedepunkt: 210 °C (1013 hPa)
Melting Pt: 77…79 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003344
CAS-Nummer: 24964-64-5
EINECS: 246-549-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR