2-Brom-p-kresol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene
AS36383G1
AS36383G5
ACTIAS36383G1EA
0
CHF
ACTIAS36383G1
ACTIAS36383G5
2-Brom-p-kresol
2-Brom-p-kresol
Formel:
C₇H₇BrO MW: 187.04 g/mol Siedepunkt: 213 Melting Pt: 26 Dichte: 1.5468 g/cm³ (25 °C) |
MDL Number:
MFCD00002151 CAS-Nummer: 6627-55-0 EINECS: 229-595-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...