2-Brom-5-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50980G1 AS50980G5
ACTIAS50980G1EA 0 CHF
ACTIAS50980G1 ACTIAS50980G5
2-Brom-5-methoxypyridin
2-Bromo-5-methoxypyridine
Formel: C₆H₆BrNO
MDL Number: MFCD07781221
CAS-Nummer: 105170-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR