2-Amino-5-brom-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

AS2615G25 AS2615G5
ACTIAS2615G25EA 0 CHF
ACTIAS2615G25 ACTIAS2615G5
2-Amino-5-brom-3-nitropyridin
2-Amino-5-brom-3-nitropyridin
Formel: C₅H₄BrN₃O₂
MW: 218,01 g/mol
Melting Pt: 215…216 °C
MDL Number: MFCD00047441
CAS-Nummer: 6945-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR