3-Brom-N,N-dimethylanilin

Lieferant: ACTIVATE SCIENTIFIC

AS35847G1 AS35847G5
ACTIAS35847G1EA 0 CHF
ACTIAS35847G1 ACTIAS35847G5
3-Brom-N,N-dimethylanilin
3-Brom-N,N-dimethylanilin
Formel: C₈H₁₀BrN
MW: 200.08 g/mol
Siedepunkt: 70 °C (0.4 mmHg)
Melting Pt: 11
Dichte: 1.3652 g/cm³ (19 °C)
MDL Number: MFCD00045020
CAS-Nummer: 16518-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR