2-Brom-4-chlorpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS50236G1
AS50236G5
ACTIAS50236G1EA
0
CHF
ACTIAS50236G1
ACTIAS50236G5
2-Brom-4-chlorpyridin
2-Bromo-4-chloropyridine
Formel:
C₅H₃BrClN |
MDL Number:
MFCD03840757 CAS-Nummer: 22918-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...