2,4-Dibrompyrimidin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS30524G1
AS30524G5
ACTIAS30524G1EA
0
CHF
ACTIAS30524G1
ACTIAS30524G5
2,4-Dibrompyrimidin
2,4-Dibromopyrimidine
Formel:
C₄H₂Br₂N₂ |
MDL Number:
MFCD06798232 CAS-Nummer: 3921-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...