Eisen(III)bromid

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS24095G100 AS24095G25
ACTIAS24095G100EA 0 CHF
ACTIAS24095G100 ACTIAS24095G25
Eisen(III)bromid
Eisen(III)bromid
Formel: Br₃Fe
MDL Number: MFCD00016082
CAS-Nummer: 10031-26-2
EINECS: 233-089-1
UN: 3260
ADR: 8,III
Merck Index: 12,04060

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR