3-Brom-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-2-picoline

AS21312G25 AS21312G5
ACTIAS21312G25EA 51 CHF
ACTIAS21312G25 ACTIAS21312G5
3-Brom-2-methylpyridin
3-Brom-2-methylpyridin
Formel: C₆H₆BrN
MW: 172,02 g/mol
MDL Number: MFCD00191224
CAS-Nummer: 38749-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR