2,4-Dimethoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50272G100 AS50272G5
ACTIAS50272G100EA 0 CHF
ACTIAS50272G100 ACTIAS50272G5
2,4-Dimethoxyphenylboronsäure
2,4-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01074590
CAS-Nummer: 133730-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR