1-Phenyl-1H-pyrazol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
1-Phenyl-1H-pyrazole, 1-Phenylpyrazole
AS34244G10
AS34244G100
ACTIAS34244G10EA
0
CHF
ACTIAS34244G10
ACTIAS34244G100
1-Phenyl-1H-pyrazol
1-Phenyl-1H-pyrazol
Formel:
C₉H₈N₂ MW: 144,18 g/mol Siedepunkt: 251 °C (1013 hPa) Melting Pt: 11 °C Dichte: 1,091 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003112 CAS-Nummer: 1126-00-7 EINECS: 214-415-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...