2-(Methylthio)phenol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Hydroxythioanisole
AS23421G100
AS23421G25
ACTIAS23421G100EA
0
CHF
ACTIAS23421G100
ACTIAS23421G25
2-(Methylthio)phenol
2-(Methylthio)phenol
Formel:
C₇H₈OS MW: 140,21 g/mol Siedepunkt: 81…82 °C (0.5 mmHg) Dichte: 1.168 g/cm³ (25 °C) |
MDL Number:
MFCD00002211 CAS-Nummer: 1073-29-6 EINECS: 214-027-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...