2-Methyl-4-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 1-Methoxy-2-methyl-4-nitrobenzene

AS21524G25 AS21524G5
ACTIAS21524G25EA 0 CHF
ACTIAS21524G25 ACTIAS21524G5
2-Methyl-4-nitroanisol
2-Methyl-4-nitroanisol
Formel: C₈H₉NO₃
MW: 167.16 g/mol
Siedepunkt: 311
Melting Pt: 96
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07787583
CAS-Nummer: 50741-92-9
EINECS: 202-763-7
UN: 2446
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR