2,4-Diamino-6-hydroxypyrimidine
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Aminoisocytosin, 2,4-Diamino-6-pyrimidinol, 2,4-Diaminopyrimidin-6-ol
AS33438G10
AS33438G100
ACTIAS33438G10EA
0
CHF
ACTIAS33438G10
ACTIAS33438G100
2,4-Diamino-6-hydroxypyrimidine
2,4-Diamino-6-hydroxypyrimidine
Formel:
C₄H₆N₄O MW: 126,12 g/mol Siedepunkt: 373 °C (1013 hPa) Melting Pt: 285…286 °C |
MDL Number:
MFCD00006098 CAS-Nummer: 56-06-4 EINECS: 200-254-4 Merck Index: 13,03001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...