2-Amino-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33431G10 AS33431G100 AS33431G5
ACTIAS33431G10EA 0 CHF
ACTIAS33431G10 ACTIAS33431G100 ACTIAS33431G5
2-Amino-3-nitropyridin
2-Amino-3-nitropyridin
Formel: C₅H₅N₃O₂
Melting Pt: 166…168 °C
Dichte: 1,54 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006314
CAS-Nummer: 4214-75-9
EINECS: 224-144-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR