4-Aminoindol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole

AS21445G1 AS21445G5
ACTIAS21445G1EA 45 CHF
ACTIAS21445G1 ACTIAS21445G5
4-Aminoindol
4-Aminoindole
Formel: C₈H₈N₂
MW: 132,16 g/mol
Melting Pt: 108 °C
MDL Number: MFCD01076559
CAS-Nummer: 5192-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR