2-Amino-3-iod-5-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Iodo-5-nitropyridin-2-amine, 3-Iodo-5-nitro-2-pyridylamine

AS30799G5 AS30799G1
ACTIAS30799G5EA 0 CHF
ACTIAS30799G5 ACTIAS30799G1
2-Amino-3-iod-5-nitropyridin
2-Amino-3-iodo-5-nitropyridine
Formel: C₅H₄IN₃O₂
MW: 265,01 g/mol
MDL Number: MFCD07368863
CAS-Nummer: 25391-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR