Guanidiniumchlorid

Lieferant: MARKET SOURCE PART PROCESS
Achtung

Synonyms: Aminomethanamidin HydrochloridCarbamidin HydrochloridGuanidin-ChloridAminomethansäureamidiniumchloridGuanidinhydrochlorid

MSPP140000279-20KGEA 0 CHF
MSPP140000279-20KG
Guanidiniumchlorid
Guanidiniumchlorid
Formel: NH₂C(=NH)NH₂·HCl
MW: 95,53 g/mol
Melting Pt: 185 °C
Dichte: 1,354 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://ch.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR