2-Fluor-3-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoropyridin-3-ol, 2-Fluoro-3-pyridinol

AS30829G1 AS30829G5
ACTIAS30829G1EA 0 CHF
ACTIAS30829G1 ACTIAS30829G5
2-Fluor-3-hydroxypyridin
2-Fluor-3-hydroxypyridin
Formel: C₅H₄FNO
MW: 113,09 g/mol
MDL Number: MFCD04112569
CAS-Nummer: 174669-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR