2-Fluor-5-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS35071G1 AS35071G5
ACTIAS35071G1EA 0 CHF
ACTIAS35071G1 ACTIAS35071G5
2-Fluor-5-iodbenzoesäure
2-Fluor-5-iodbenzoesäure
Formel: C₇H₄FIO₂
MW: 266,01 g/mol
MDL Number: MFCD03094517
CAS-Nummer: 124700-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR