2-Fluor-3-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS50172G1
AS50172G5
ACTIAS50172G1EA
0
CHF
ACTIAS50172G1
ACTIAS50172G5
2-Fluor-3-iodpyridin
2-Fluor-3-iodpyridin
Formel:
C₅H₃FIN MW: 222,99 g/mol |
MDL Number:
MFCD03095287 CAS-Nummer: 113975-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...