4-Fluor-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31099G1 AS31099G5
ACTIAS31099G1EA 0 CHF
ACTIAS31099G1 ACTIAS31099G5
4-Fluor-2-nitrobenzaldehyd
4-Fluoro-2-nitrobenzaldehyde
Formel: C₇H₄FNO₃
MDL Number: MFCD01666439
CAS-Nummer: 2923-96-8
EINECS: 220-884-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR