4-Nitrosalicylsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Hydroxy-4-nitrobenzoic acid
AS40473G1
AS40473G5
ACTIAS40473G1EA
0
CHF
ACTIAS40473G1
ACTIAS40473G5
4-Nitrosalicylsäure
4-Nitrosalicylsäure
Formel:
C₇H₅NO₅ MW: 183.12 g/mol |
MDL Number:
MFCD00071505 CAS-Nummer: 619-19-2 EINECS: 210-584-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...