2-Nitrozimtsäure
Lieferant: ACTIVATE SCIENTIFIC
AS23299G100
AS23299G25
ACTIAS23299G100EA
0
CHF
ACTIAS23299G100
ACTIAS23299G25
2-Nitrozimtsäure
2-Nitrozimtsäure
Formel:
C₉H₇NO₄ |
CAS-Nummer:
612-41-9 EINECS: 210-309-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...