3-Methoxyanthranilsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid
AS33536G1
AS33536G5
ACTIAS33536G1EA
0
CHF
ACTIAS33536G1
ACTIAS33536G5
3-Methoxyanthranilsäure
3-Methoxyanthranilsäure
Formel:
C₈H₉NO₃ Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00075178 CAS-Nummer: 3177-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...