2-Methylzimtsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methylcinnamic Acid, Predominantly trans
AS24409G100
AS24409G25
ACTIAS24409G100EA
0
CHF
ACTIAS24409G100
ACTIAS24409G25
2-Methylzimtsäure
2-Methylzimtsäure
Formel:
C₁₀H₁₀O₂ MW: 162.19 g/mol Siedepunkt: 303 Melting Pt: 175…176 °C |
MDL Number:
MFCD00016841 CAS-Nummer: 2373-76-4 EINECS: 219-150-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...