D(-)-Glutamin
Lieferant: ACTIVATE SCIENTIFIC
AS34951G1
AS34951G10
AS34951G5
ACTIAS34951G1EA
0
CHF
ACTIAS34951G1
ACTIAS34951G10
ACTIAS34951G5
D(-)-Glutamin
D(-)-Glutamin
Formel:
C₅H₁₀N₂O₃ |
MDL Number:
MFCD00065607 CAS-Nummer: 5959-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...