2'-Nitroacetophenon
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
AS20737G100
AS20737G25
ACTIAS20737G100EA
95
CHF
ACTIAS20737G100
ACTIAS20737G25
2'-Nitroacetophenon
2'-Nitroacetophenon
Formel:
C₈H₇NO₃ MW: 165,15 g/mol Siedepunkt: 286 °C (1013 hPa) Melting Pt: 23 °C Dichte: 1,24 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007144 CAS-Nummer: 577-59-3 EINECS: 209-414-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...