2-Chlor-6-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS1574G1 AS1574G5
ACTIAS1574G1EA 0 CHF
ACTIAS1574G1 ACTIAS1574G5
2-Chlor-6-nitropyridin
2-Chloro-6-nitropyridine
Formel: C₅H₃ClN₂O₂
MW: 158,54 g/mol
MDL Number: MFCD04114207
CAS-Nummer: 94166-64-0
EINECS: 303-361-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR